Learn More
SP Genevac™ miVac™ 2 Stacker for swing rotor
Product Code. 31723011
Change view
Click to view available options
Type:
Adapter Set
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
31723011 Adapter Set Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31723011 Supplier SP Genevac™ Supplier No. BRSSTK00016
 Restricted Item
Estimated Shipment: 23-11-2026
Allows user to stack up to 5 shallow well plate per swing.
  • This Stacker accessory for miVac allows for up to 5 shallow well plate per swing.
Specifications
Type Adapter Set