Learn More
Mitoquinone mesylate, MedChemExpress
Product Code. 16449476
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449476 50 mg 50mg
16495517 5 mg 5mg
16452526 25 mg 25mg
16402788 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16449476 Supplier Medchem Express Supplier No. HY100116A50mg
MedChemExpress Mitoquinone mesylate is a TPP-based, mitochondrially targeted antioxidant in order to protect against oxidative damage.
Mitoquinone mesylate is a TPP-based, mitochondrially targeted antioxidant in order to protect against oxidative damage.
Specifications
Chemical Name or Material Mitoquinone mesylate
CAS 845959-50-4
Color Brown
Purity Grade Notes Research
Molecular Formula C38H47O7PS
Quantity 50 mg
Synonym MitoQ mesylateMitoQ10 mesylate
Solubility Information DMSO : 50 mg/mL (73.66 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (12.27 mM; ultrasonic and warming and heat to 60°C)
SMILES [O-]S(=O)(C)=O.O=C(C(CCCCCCCCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)=C4C)C(OC)=C(OC)C4=O
Molecular Weight (g/mol) 678.81
Formula Weight 678.81
Percent Purity 98.0%
Grade Research
Physical Form Oil
For Use With (Application) Cancer-programmed cell death