missing translation for 'onlineSavingsMsg'
Learn More

Mirk-IN-1, MedChemExpress

Product Code. p-200099083
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465388 10 mg 10mg
16451577 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16465388 Supplier Medchem Express Supplier No. HY12838
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Mirk-IN-1 is a potent inhibitor of Dyrk1B(Mirk kianse) and Dyrk1A with IC50 of 68±48 nM and 22±8 nM respectively.

Mirk-IN-1 is a potent inhibitor of Dyrk1B(Mirk kianse) and Dyrk1A with IC50 of 68±48 nM and 22±8 nM respectively.

Specifications

Chemical Name or Material Mirk-IN-1
CAS 1386979-55-0
Color White
Purity Grade Notes Research
Molecular Formula C23H17Cl2N5O4
Quantity 10 mg
Synonym Dyrk1B/A-IN-1
Solubility Information DMSO : 5 mg/mL (10.03 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES ClC1=CC=C(C(NCC2=CC(Cl)=CC=C2)=O)C=C1NC(C3=CC4=CN=C(OC)N=C4N=C3O)=O
Molecular Weight (g/mol) 498.32
Formula Weight 498.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.