Learn More
Miramistin, TRC
Product Code. 17940075
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17940075 500 mg 500mg
17907704 250 mg 250mg
17939334 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17940075 Supplier Toronto Research Chemicals Supplier No. TRCM365035500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15809-19-5
Molecular Formula C26 H47 N2 O . Cl
Molecular Weight (g/mol) 439.12
Synonym N,N-Dimethyl-N-[3-[(1-oxotetradecyl)amino]propyl]-benzenemethanaminium Chloride, Benzyldimethyl(3-tetradecamidopropyl)ammonium Chloride, Benzyldimethyl[3-(myristoylamino)propyl]ammonium Chloride, Myramistin, Benzyldimethyl(3-myristamidopropyl)-ammonium Chloride, DT 2, DT 2 (surfactant);
IUPAC Name benzyl-dimethyl-[3-(tetradecanoylamino)propyl]ammonium;chloride
SMILES [Cl-].CCCCCCCCCCCCCC(=O)NCCC[N+](C)(C)Cc1ccccc1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C26H46N2O.ClH/c1-4-5-6-7-8-9-10-11-12-13-17-21-26(29)27-22-18-23-28(2,3)24-25-19-15-14-16-20-25;/h14-16,19-20H,4-13,17-18,21-24H2,1-3H3;1H
Formula Weight 438.3377
Chemical Name or Material Miramistin