missing translation for 'onlineSavingsMsg'
Learn More

Ministry of Agriculture Veterinary Drug Mixture 348 10 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer

Product Code. 30317355
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317355 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317355 Supplier Dr. Ehrenstorfer Supplier No. DREA50000348WL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Danofloxacin∣Difloxacin hydrochloride∣Enrofloxacin∣Lomefloxacin∣Marbofloxacin∣Ofloxacin∣Pefloxacin∣Sulfabenzamide∣Sulfacetamide∣Sulfachloropyrazine∣Sulfachlorpyridazine Sodium∣Sulfadiazine∣Sulfadimethoxine∣Sulfadoxine∣Sulfaguanidine∣Sulfamerazine∣Sulfamethoxazole∣Sulfamonomethoxine∣Sulfanilamide∣Sulfaphenazole∣Sulfapyridine∣Sulfaquinoxaline∣Sulfisomidine, Sodium∣Sulfisoxazole∣Trimethoprim
CAS 112398-08-0, 91296-86-5, 93106-60-6, 98079-51-7, 115550-35-1, 82419-36-1
Analyte or Component Names Danofloxacin∣Difloxacin hydrochloride∣Enrofloxacin∣Lomefloxacin∣Marbofloxacin∣Ofloxacin∣Pefloxacin∣Sulfabenzamide∣Sulfacetamide∣Sulfachloropyrazine∣Sulfachlorpyridazine Sodium∣Sulfadiazine∣Sulfadimethoxine∣Sulfadoxine∣Sulfaguanidine∣Sulfamerazine∣Sulfamethoxazole∣Sulfamonomethoxine∣Sulfanilamide∣Sulfaphenazole∣Sulfapyridine∣Sulfaquinoxaline∣Sulfisomidine, Sodium∣Sulfisoxazole∣Trimethoprim
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile/Water
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Danofloxacin 10 μg/mL∣Difloxacin hydrochloride 10 μg/mL∣Enrofloxacin 10 μg/mL∣Lomefloxacin 10 μg/mL∣Marbofloxacin 10 μg/mL∣Ofloxacin 10 μg/mL∣Pefloxacin 10 μg/mL∣Sulfabenzamide 10 μg/mL∣Sulfacetamide 10 μg/mL∣Sulfachloropyrazine 10 μg/mL∣Sulfachlorpyridazine Sodium 10 μg/mL∣Sulfadiazine 10 μg/mL∣Sulfadimethoxine 10 μg/mL∣Sulfadoxine 10 μg/mL∣Sulfaguanidine 10 μg/mL∣Sulfamerazine 10 μg/mL∣Sulfamethoxazole 10 μg/mL∣Sulfamonomethoxine 10 μg/mL∣Sulfanilamide 10 μg/mL∣Sulfaphenazole 10 μg/mL∣Sulfapyridine 10 μg/mL∣Sulfaquinoxaline 10 μg/mL∣Sulfisomidine, Sodium 10 μg/mL∣Sulfisoxazole 10 μg/mL∣Trimethoprim 10 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.