missing translation for 'onlineSavingsMsg'
Learn More

Minioxidil, 98.2%, MP Biomedicals™

Product Code. 11474911 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474911 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474911 Supplier MP Biomedicals Supplier No. 0215571980
  • ONLINE EXCLUSIVE PRICE
    132.00 EUR
    124.95 EUR / 100mg
    Save 7.05 EUR
    5% Off
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 38304-91-5
Color White
Assay Percent Range 0.982
Molecular Formula C9H15N5O
Quantity 100 mg
Synonym minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
InChI Key ZIMGGGWCDYVHOY-UHFFFAOYSA-N
SMILES C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O
IUPAC Name 3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
Molecular Weight (g/mol) 209.253
PubChem CID 4201
Formula Weight 209.3
Physical Form Crystalline Powder
Chemical Name or Material Minioxidil
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.