Learn More
Bryson Packaging™ Minigrip™ Polyethylene Write-on Bags
Artikelnummer. 11597863
Ansicht ändern
Click to view available options
Width (Metric):
57 mm
60 mm
90 mm
115 mm
125 mm
150 mm
230 mm
254 mm
279 mm
Height (Metric):
60 mm
76 mm
115 mm
190 mm
230 mm
325 mm
356 mm
406 mm
Packungsgröße:
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Width (Metric) Height (Metric) Packungsgröße
11597863 125 mm 190 mm Pack of 100
11388193 115 mm 115 mm Pack of 100
11998267 230 mm 325 mm Pack of 100
11368203 254 mm 356 mm Pack of 100
11378203 279 mm 406 mm Pack of 100
11358203 57 mm 76 mm Pack of 100
11378193 60 mm 60 mm Pack of 100
11587863 90 mm 115 mm Pack of 100
11988267 150 mm 230 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Artikelnummer. 11597863 Lieferant Bryson Packaging™ Lieferanten-Nr. BAJ340071T
In Stock
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen
Store and send samples and stationary in resealable bags. Bryson Packaging™ Minigrip™ Polyethylene Write-on Bags are clear polyethylene bags with three white bars for labelling. Square and rectangular bags are available in a range of sizes. 100 and 1,000 packs.
  • Clear polyethylene bags with Grip Seal resealable strips
  • Three generously sized write-on labelling bars for easy identification and organization
  • Ideal for storing stationery and sending samples
  • Available in a range of sizes to meet storage needs
Spezifikation
Description Resealable Write-On Bag
Color Clear with White Bars
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Quantity 100 Pk.
Height (Metric) 190 mm
Width (Metric) 125 mm
Thickness (Metric) 50 μm