missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Small, mono-disperse beads for extremely high resolution purifications. non porous beads contribute to high resolution but give lower capacity than MonoBeads
- Analytical and micro-preparative applications with choice of both anion and cation exchangers
- High chemical and physical stability
- Excellent reproducibility and durability
- Tricorn high-performance columns (4.6/50 PE) prepacked with MiniBeads
- PC columns designed for micropurification
Precision column holders may be required for use on high performance chromatography systems such as KTA design. For optimum results, it is important to check the compatibility of the column and the system specifications.
Specifications
Specifications
| Buffer | All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid |
| Product Type | Mini S Ion Exchange Column |
| Bed Volume | 0.8 mL |
| Diameter (English) Inner | 0.18 in. |
| Diameter (Metric) Inner | 4.6 mm |
| Quantity | Each |
| Height (English) | 1.96 in. |
| Height (Metric) | 50 mm |
| Flow Rate | 0.5 to 2 mL/min. |
| Includes | Valco fittings |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?