Learn More
Fisherbrand™ Mini UV Sterilization Cabinet with Timer
Product Code. 15582496
Change view
Click to view available options
Includes:
No tray
Tray
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15582496 Tray Each
15572496 No tray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15582496
  • ONLINE EXCLUSIVE PRICE
    1675.00 EUR
    1629.11 EUR / Pack of 1
    Save 45.89 EUR
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-02-2027
Limits PCR reaction contamination. Fisherbrand™ Mini UV Sterilization Cabinet with Timer provides a convenient area for setting up PCR reactions in a nucleic acid-free environment.
  • UV lights on the cabinets denature nucleic acids in 5 to 30 minutes making them unsuitable for amplification.
  • Incorporates safety features to prevent user exposure to UV light.
  • The UV lights are timer controlled
  • Safety switches on the cabinets doors turn off the UV lights when opened.
  • Shields beta radioactive emissions
  • Specifications
    Lighting Type 4 UV Lights, 1 White
    Depth (Metric) 350 mm
    Height (Metric) 510 mm
    Includes Tray
    Wattage 5 x 15 W
    Weight (Metric) 12 kg
    Width (Metric) 560 mm