Learn More
Fisherbrand™ Mini Electrophoresis System
Product Code. 15530340 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Gel Dimensions:
One: 105 x 60 mm, Two: 50 x 60 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gel Dimensions Unit Size
15530340 One: 105 x 60 mm, Two: 50 x 60 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15530340
  • ONLINE EXCLUSIVE PRICE
    685.00 EUR
    500.92 EUR / Pack of 1
    Save 184.08 EUR
    26% Off
View all promotions
Estimated Shipment: 13-01-2027
Add to basket
Add to basket
Pour gel, load contrast strips, and press start. Fisherbrand™ mini Electrophoresis System is ideal for personal use, small laboratories or the classroom and includes gel take, casting sets and power supply.
  • Includes gel box power supply and two casting sets
  • Quick and easy gel casting
  • No clamps or spacers required
  • Compact design
Specifications
Timer 0 to 99 min. or continuous
Gel Dimensions One: 105 x 60 mm, Two: 50 x 60 mm
Includes Safety lid interlock, two casting sets
Volume (Metric) Buffer 230 mL
Dimensions (L x W x H) 190 x 130 x 55 mm
Electrical Requirements 115/230 V 50/60 Hz
Frequency 50/60 Hz