Learn More
ITW Chemische Produkte™ Mini AlphaMop™ AlphaWipe™ Replacement Mop Covers
Product Code. 15664987 Shop All Texwipe Products
Change view
Click to view available options
Material:
Polyester
Unit Size:
Pack of 150
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15664987 Polyester Pack of 150
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15664987 Supplier ITW Chemische Produkte™ Supplier No. TX7114E
  • ONLINE EXCLUSIVE PRICE
    440.00 EUR
    375.99 EUR / Pack of 150
    Save 64.01 EUR
    14% Off
Estimated Shipment: 08-12-2026
Designed for use with the AlphaMop™ Isolator Cleaning Tool. Mini AlphaMop™ AlphaWipe™ Replacement Mop Covers are double bagged and are made of AlphaWipe™ polyester for effective cleaning with low contamination.
  • Made of AlphaWipe™ polyester for effective cleaning and low contamination
  • Cleanroom laundered for use in sterile and aseptic applications
  • Designed to slip on and off easily, yet remain secure while in use
  • Replacement mop pads available separately
Specifications
Product Type Mop Cover
For Use With (Equipment) AlphaMop Isolator Cleaning Tool
Description Double Bagged
Material Polyester