Learn More
Milrinone, LoGiCal
Product Code. 30458143
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458143 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30458143 Supplier LoGiCal Supplier No. LGCFOR1539.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Milrinone
CAS 78415-72-2
Analyte or Component Names Milrinone
Shipping Condition Room Temperature
Molecular Formula C12 H9 N3 O
Certifications/Compliance ISO 9001
Synonym Milrinone, [3,4'-Bipyridine]-5-carbonitrile, 1,6-dihydro-2-methyl-6-oxo-, 1,6-Dihydro-2-methyl-6-oxo-(3,4'-bipyridine)-5-carbonitrile, Corotrop, Corotrope, Milrinone, Primacor IV, Win 47203
InChI Formula InChI=1S/C12H9N3O/c1-8-11(9-2-4-14-5-3-9)6-10(7-13)12(16)15-8/h2-6H,1H3,(H,15,16)
Solution Type Neat
SMILES CC1=C(C=C(C#N)C(=O)N1)c2ccncc2
IUPAC Name 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
Industry Type Clinical Testing
Molecular Weight (g/mol) 211.22
Quantity 10 mg
Formula Weight 211.0746 g/mol