Learn More
Melford Millers LB Broth Granulated
Product Code. 30018428
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018428 1 kg 1kg
30018430 5 kg 5kg
30018429 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30018428 Supplier Melford Supplier No. L244001000.0
For propagation and maintenance of E.
For propagation and maintenance of E. Coli. High Salt Formula. Molecular Biology Grade.
Dissolve 25 grams in 1 liter distilled water and adjust the pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes. Material specially granulated to reduce dust and increase solubility rate.
Ingredients:
Tryptone:10 g/l
Sodium Chloride: 10 g/l
Yeast Extract: 5 g/l
Specifications
Quantity 1 kg