missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Mifamurtide
Specifications
Specifications
| Purity | 98.0% |
| CAS | 83461-56-7 |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | Cancer-programmed cell death |
| Molecular Weight (g/mol) | 1237.5 g/mol |
| Product Type | Reference compound |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 100 mg/mL (80.81 mM∣Need ultrasonic) |
| Synonym | MTP-PE∣ L-MTP-PE∣ CGP 19835 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?