Learn More
IKA™ Microstar 7.5 Control
GREENER_CHOICE
Product Code. 16238611 Shop All IKA Products
Change view
Click to view available options
Speed:
2000 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed unitSize
16238611 2000 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16238611 Supplier IKA™ Supplier No. 0025001984
Only null left
Add to basket
Add to basket
High-tech overhead stirrer features a compact design, making it ideal for special applications.
  • Hardened glass enclosed, fast response display for maximum visibility and chemical resistance
  • State-of-art vibration sensors detect deviations from permissible thresholds and automatically stop the process
  • Clear display for all essential information at a glance
  • Integrated timer / counter for the control of kinetic sensitive reactions and reminders
  • Viscosities up to 4000 mPas and volume of up to 5 L
  • Continuously adjustable speed between 30 to 2000 rpm
  • USB interface, e.g. for documenting parameters using labworldsoft™ or installing firmware updates
  • Intuitive and simple handling; touch-sensitive surface for long service life
  • Temperature reading on display
  • Chemically resistant housing
  • Key lock function
  • Microprocessor-controlled speed governor for constant rotational speed, even with changes in viscosity
TRUSTED_SUSTAINABILITY
Specifications
Speed 2000 rpm
Capacity 5 L
Display LCD
Voltage 100/240 V
Wattage 32 w
Diameter (Metric) Shaft 0.5 to 8 mm
Temperature Range (Metric) -10°C to +350°C
Torque 7.5 Ncm
Viscosity Max. 4000 mPas