Learn More
Applied Biosystems™ MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0, with all library kits
Shop All Applied Biosystems ProductsDescription
- Allows users to easily identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library
- Automatically identifies unknown specimens
- Features a validated library of over 11,000 microbial sequences (with all updated library kits)
- MicroSEQ™ ID sequence libraries consist of bacterial libraries of 500-bp 16S rDNA sequences and over 9500 validated entries
- Extensive coverage of gram-negative non-fermenters, Bacillus, coryneforms, mycobacteria, and Staphylococcus
- Libraries contain only correctly identified organisms and quality-controlled sequences derived from type strains of preeminent culture collections
- Supports sequence polymorphisms and includes bacterial Category A biothreat agents
- Run a plate on the SeqStudio™ Genetic Analyzer directly via the MicroSEQ™ ID Software
- MicroSEQ™ system includes security, audit, and e-signature features
- Qualification and validation support available
Spécification
Spécification
| Product Line | MicroSEQ ID Software |
| Description | Includes digital download letter, license code kit, library kits (USB media and license code kits for 2018 Fungal Library, 2019 16S 500 Library, 2019 16s 500 Supplemental Library, and 16s Full Gene Library) |
| For Use With (Application) | Microbial risk mitigation, identification |
| For Use With (Equipment) | QST SeqStudio™ Instrument with SAE |
| Libraries | Included |
| License | Included |
| Quantity | 1 Pk. |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?