Learn More
Hirschmann™ Microscopic Slide with Cut Edges
Product Code. 10049700
Change view
Click to view available options
Quantity:
1000/Pk.
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10049700 1000/Pk. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10049700 Supplier Hirschmann™ Supplier No. 8200103
  • ONLINE EXCLUSIVE PRICE
    81.00 EUR
    69.30 EUR / Pack of 1000
    Save 11.70 EUR
    14% Off
 Restricted Item
Estimated Shipment: 21-12-2026
An excellent choice for microscopy applications in the laboratory.
  • 76 x 26 mm
  • Thickness from 0.95-1.05 mm
  • Extrawhite in color
  • With LOT number on packaging
  • Labelled with: usable until, CE pursuant to IVD 98/79 EC
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
Edges Cut
Thickness (Metric) 0.95 to 1.05 mm
Type Microscope Slide
Color Extra White
Certifications/Compliance CE pursuant to IVD 98/79 EC
For Use With (Application) Microscopy
Quantity 1000/Pk.
Dimensions (L x W) 76 x 26 mm