Learn More
Thermo Scientific™ Aluminum Support, tube/plate rack, 96-well
Product Code. 10403420 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 rack
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10403420 1 rack 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10403420 Supplier Thermo Scientific™ Supplier No. PRK1963
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 06-11-2026
Thermo Scientific™ aluminum support rack can be used for sample preparation, heat-sealing and automatic dispensing.
Perform sample preparation, heat-sealing and automatic dispensing using Thermo Scientific™ aluminum support rack.
Available in 96- and 384-well format.
  • For heat-sealing and automated dispensing
  • Industry-standard footprint fits in robotic workstations
  • Can be heated or cooled to assist with sample preparation
Includes:
Recommended for:
Sample handling, sample preparation, heat-sealing, automated dispensing
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Holds Microplates (-well)
Material Aluminum
Shape Rectangular
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Quantity 1 rack
Product Type Microplate Rack