Learn More
Socorex™ Micropipette Rack for Qualitix™ Tipfill Refill System
Product Code. 15770649
Change view
Click to view available options
Holds:
Micropipette Tips (10μL), Micropipette Tips (200μL )
Micropipette Tips (1000μL)
No. per Pack:
6
10
Unit Size:
Pack of 10
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds No. per Pack unitSize
15770649 Micropipette Tips (10μL), Micropipette Tips (200μL ) 10 Pack of 10
15790649 Micropipette Tips (1000μL) 6 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15770649 Supplier Socorex™ Supplier No. 3070.0210ER
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
Reusable, reclosable, and more space-saving than conventional racks. Socorex™ Micropipette Rack for Qualitix™ Tipfill Refill System readily accepts micropipette refill packs and has smooth, rounded edges and an ergonomic snap latch that won't catch gloves.
  • Accepts Qualitix™ Tipfill Refill System refill packs.
  • Hinged lid.
  • Smooth, rounded edges.
  • Ergonomic snap latch that won't catch gloves.
  • Transparent lid.
  • Reusable.
Specifications
Closure Included Yes
Autoclavable Autoclavable
Color Natural
Holds Micropipette Tips (10μL), Micropipette Tips (200μL )
Stackable Yes
For Use With (Equipment) Qualitix™Tipfill Refill System
No. per Case 10
No. per Pack 10
Array 96