Learn More
Micronova™ PharmaPly™ Mop Head
BIOPHARMA
Product Code. 11906875 Shop All Micronova Products
Change view
Click to view available options
Length (English):
15 in.
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
11906875 15 in. Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11906875 Supplier Micronova™ Supplier No. PPM4MM15
  • ONLINE EXCLUSIVE PRICE
    406.00 EUR
    376.12 EUR / Pack of 24
    Save 29.88 EUR
    7% Off
 Restricted Item
Estimated Shipment: 12-04-2027
Ideal for covering large surface areas quickly and efficiently.
  • Polyester/rayon blend mop head for cleaning/disinfecting pharmaceutical support areas, hospitals and laboratories
  • Designed for use with MicroMop™ adapter and handle (sold separately)
  • 15 in. long (38cm)
Specifications
Product Type Cleanroom Mop
Color Code White
Length (English) 15 in.
Sterility Sterile
Autoclavable Autoclavable
Width (English) 6 in.
Disposable Reusable
For Use With (Application) Used for Covering Large Surface Areas Quickly and Efficiently, Floor
Color-coded No
Material Polyester, Rayon