Learn More
Ansell Edmont™ Microgard™ 3000 Cape Hood with Visor Opening
Product Code. 15566604 Shop All Ansell Products
Product Code. 15566604 Supplier Ansell Edmont™ Supplier No. YE30W0050800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Protects head and shoulders from inorganic chemicals. Ansell Edmont™ Microgard™ 3000 Cape Hood with Visor Opening should also be worn with a protective face shield.
  • Multi-layer barrier laminate
  • Balaclava style with visor to face opening
  • Drapes over part of the shoulders
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hood with Visor
Size Universal
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Design Type Balaclava style with visor opening
Seam Type Ultrasonically welded
Certifications/Compliance Category I PPE according to European Directive 89/686/EEC
Quantity 30/Pk.
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.