Learn More
Fisherbrand™ Microcentrifuge Tubes with Locking Snap Cap
Product Code. 15432545 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Natural
Sterility:
Non-sterile
Capacity (Metric):
0.5 mL
1.5 mL
2.0 mL
Max. RCF:
25,000 x g
26,000 x g
30,000 x g
Unit Size:
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Capacity (Metric) Max. RCF unitSize
15432545 Natural Non-sterile 1.5 mL 26,000 x g Pack of 500
15442545 Natural Non-sterile 2.0 mL 25,000 x g Pack of 500
15422545 Natural Non-sterile 0.5 mL 30,000 x g Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15432545
In Stock
Add to basket
Add to basket
Polypropylene microcentrifuge tubes are DNase/RNase and pyrogen free
  • DNase/RNase and pyrogen free
  • Autoclavable and withstand temperatures to -86°C
  • Graduated tubes available in three sizes - 0.5mL, 1.5mL and 2.0mL
Specifications
Cap Style Locking snap cap
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Natural
Material Polypropylene
Sterility Non-sterile
Graduated Yes
Max. RCF 26,000 x g
Diameter (Metric) 13 mm
Length (Metric) 39 mm