Learn More
BRAND™ Microcentrifuge Tubes
Product Code. 30243949 Shop All Brand Products
Change view
Click to view available options
Color:
Transparent
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243949 Transparent Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30243949 Supplier BRAND™ Supplier No. 780555
  • ONLINE EXCLUSIVE PRICE
    269.00 EUR
    240.64 EUR / Pack of 250
    Save 28.36 EUR
    10% Off
Estimated Shipment: 29-12-2026
High cleanliness for demanding applications, with tight sealing snap lid which can be opened and closed with one hand.
Disposable microtubes made from high-quality PP. Choose between 1.5 and 5 mL nominal capacity. The quality level BIO-CERT™ PCR QUALITY ensures the required cleanliness for PCR-setups, storage of PCR samples and other applications with high cleanliness requirements.
Comes with an easy-to-open snap-on lid. The disposable microtubes can be centrifuged up to the maximum RCF value stated and autoclaved at 121°C (2 bar) according to DIN EN 285. For a reading of the approximate volume, they have an easy-to-read graduation. For easy identification of samples, different colors are available.
BIO-CERT™ PCR QUALITY: Free of human DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Capacity (Metric) 5 mL
Color Transparent
Material Polypropylene
Sterility Non-sterile
Graduated Yes
Closure Type Attached Snap Cap
Bottom Shape Conical (Starfoot)
Shape Cylindrical
Product Type Microcentrifuge Tube