Learn More
SI Analytics™ Micro-Ubbelohde Viscometers (DIN)
Micro-Ubbelohde viscometers are calibrated for manual measurements
Brand: SI Analytics™ 285401017
Additional Details : Weight : 0.30000kg
Description
- Calibrated for manual measurement
- DIN
- Type No. 536 13
- Capillary No. Ic
- Constant K = 0.03
Specifications
Ubbelohde Micro Viscometer |