Learn More
Mevinphos-d6 (Major), TRC
Specifications
Specifications
| Chemical Name or Material | Mevinphos D6 |
| CAS | 2470235-45-9 |
| Molecular Formula | C7 D6 H7 O6 P |
| Quantity | 25 mg |
| Synonym | 2-Butenoic acid, 3-[(dimethoxy-D6-phosphinyl)oxy]-, methyl ester, Mevinphos D6 (P-dimethoxy D6) |
| InChI Formula | InChI=1S/C7H13O6P/c1-6(5-7(8)10-2)13-14(9,11-3)12-4/h5H,1-4H3/i3D3,4D3 |
| SMILES | [2H]C([2H])([2H])OP(=O)(OC(=CC(=O)OC)C)OC([2H])([2H])[2H] |
| IUPAC Name | methyl 3-[bis(trideuteriomethoxy)phosphoryloxy]but-2-enoate |
| Molecular Weight (g/mol) | 230.19 |
| Formula Weight | 230.0826 |