Learn More
Mettler Toledo™ DL31 Karl Fischer™ Titrator Replacement Parts: Electrodes
Product Code. 12734436 Shop All Mettler Toledo Products
Change view
Click to view available options
Type:
Combined Platinum Ring Electrode
Combined Silver Ring Electrode
For Use With (Application):
For argentometric titrations
For redox titrations
Temperature (English) Range:
0°F to 176°F
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Application) Temperature (English) Range Unit Size
12734436 Combined Platinum Ring Electrode For redox titrations 0°F to 176°F Each
10380143 Combined Silver Ring Electrode For argentometric titrations 0°F to 176°F Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12734436 Supplier Mettler Toledo™ Supplier No. 89598
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replacement parts for DL31 Karl Fischer titrators.
Combined platinum ring redox electrode with a ceramic frit for redox titrations with change of the pH value.
TRUSTED_SUSTAINABILITY
Specifications
Type Combined Platinum Ring Electrode
For Use With (Application) For redox titrations
Includes Ceramic frit
Length (English) 5.8 in.
Length (Metric) 149.5 mm
Temperature (English) Range 0°F to 176°F
Temperature (Metric) Range 0°C to 80°C