Learn More
Mettler Toledo™ Accessory for Titrator: Titration Vessels
Product Code. 10354510 Shop All Mettler Toledo Products
Change view
Click to view available options
Type:
250 mL Glass Titration Vessel
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10354510 250 mL Glass Titration Vessel Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10354510 Supplier Mettler Toledo™ Supplier No. 00023515
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
The right beaker for the right sample
  • 09-313-151D: 250mL glass beakers for attachment directly to titration stand (set of 10)
  • 09-313-151Q: 80mL glass beakers (set of 20)
TRUSTED_SUSTAINABILITY
Specifications
Type 250 mL Glass Titration Vessel
For Use With (Application) Titration
Material Glass
Quantity 10 Pk.
Volume (Metric) 250 mL
For Use With (Equipment) Titrator