missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole MP Biomedicals

Product Code. 11434901 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434901 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434901 Supplier MP Biomedicals Supplier No. 0215571005
  • ONLINE EXCLUSIVE PRICE
    50.70 EUR
    48.30 EUR / 5g
    Save 2.40 EUR
    4% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibiotic agent that suppresses protozoans and anaerobes with superior tissue penetration.

Specifications

CAS 443-48-1
Molecular Formula C6H9N3O3
Quantity 5 g
Synonym metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
InChI Key VAOCPAMSLUNLGC-UHFFFAOYSA-N
SMILES CC1=NC=C(N1CCO)[N+](=O)[O-]
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)ethanol
Molecular Weight (g/mol) 171.156
PubChem CID 4173
ChEBI CHEBI:6909
Formula Weight 171.2
Grade Not Available
Physical Form Powder
Chemical Name or Material Metronidazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.