missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole, 99.32%, MP Biomedicals™

Product Code. 11454901 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454901 100 g 100g
11444901 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454901 Supplier MP Biomedicals Supplier No. 0215571080
  • ONLINE EXCLUSIVE PRICE
    405.00 EUR
    385.35 EUR / 100g
    Save 19.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 443-48-1 | C6H9N3O3 | 171.156 g/mol

Chemical Identifiers

CAS 443-48-1
Molecular Formula C6H9N3O3
Molecular Weight (g/mol) 171.156
InChI Key VAOCPAMSLUNLGC-UHFFFAOYSA-N
Synonym metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
PubChem CID 4173
ChEBI CHEBI:6909
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)ethanol
SMILES CC1=NC=C(N1CCO)[N+](=O)[O-]

Specifications

Color White
Quantity 100 g
Formula Weight 171.2
Percent Purity 0.9932
Grade Not Available
Physical Form Powder
Chemical Name or Material Metronidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.