Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Product Code. 15926591
Change view
Click to view available options
Dimensions (L x W):
35.5 x 121.9 cm (14 x 48 in.)
35.5 x 182.9 cm (14 x 72 in.)
45.7 x 121.9 cm (18 x 48 in.)
45.7 x 183 cm (18 x 72 in.)
45.7 x 61 cm (18 x 24 in.)
45.7 x 91.4 cm (18 x 36 in.)
53 x 106.6 cm (21 x 42 in.)
53 x 21.9 cm (21 x 48 in.)
53 x 37 cm (21 x 54 in.)
53 x 60 cm (21 x 24 in.)
53 x 76 cm (21 x 30 in.)
61 x 106.6 cm (24 x 42 in.)
61 x 121.9 cm (24 x 48 in.)
61 x 137 cm (24 x 54 in.)
61 x 152.4 cm (24 x 60 in.)
61 x 91.4 cm (24 x 36 in.)
76 x 121.9 cm (30 x 48 in.)
76 x 182.9 cm (30 x 72 in.)
76 x 91.4 cm (30 x 36 in.)
91 x 152.4 cm (36 x 60 in.)
91 x 182.9 cm (36 x 72 in.)
91 x 91 cm (36 x 36 in.)
Depth (Metric):
355 mm
457 mm
530 mm
610 mm
760 mm
910 mm
Unit Size:
Each
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Depth (Metric) Unit Size
15926591 45.7 x 61 cm (18 x 24 in.) 457 mm Each
15946611 45.7 x 183 cm (18 x 72 in.) 457 mm Each
15966681 76 x 121.9 cm (30 x 48 in.) 760 mm Each
15946681 76 x 91.4 cm (30 x 36 in.) 760 mm Each
15916671 61 x 121.9 cm (24 x 48 in.) 610 mm Each
15976671 61 x 152.4 cm (24 x 60 in.) 610 mm Each
15966601 45.7 x 121.9 cm (18 x 48 in.) 457 mm Each
15936661 61 x 91.4 cm (24 x 36 in.) 610 mm Each
15906601 45.7 x 91.4 cm (18 x 36 in.) 457 mm Each
15906581 35.5 x 121.9 cm (14 x 48 in.) 355 mm Each
15986581 35.5 x 182.9 cm (14 x 72 in.) 355 mm Each
15996611 53 x 60 cm (21 x 24 in.) 530 mm Each
15986631 53 x 21.9 cm (21 x 48 in.) 530 mm Each
15946621 53 x 76 cm (21 x 30 in.) 530 mm Each
15936631 53 x 106.6 cm (21 x 42 in.) 530 mm Each
15916641 53 x 37 cm (21 x 54 in.) 530 mm Each
15986661 61 x 106.6 cm (24 x 42 in.) 610 mm Each
15946671 61 x 137 cm (24 x 54 in.) 610 mm Each
15926691 76 x 182.9 cm (30 x 72 in.) 760 mm Each
15916701 91 x 91 cm (36 x 36 in.) 910 mm Each
15986701 91 x 182.9 cm (36 x 72 in.) 910 mm Each
15956701 91 x 152.4 cm (36 x 60 in.) 910 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Product Code. 15926591 Supplier Metro™ Supplier No. 1824NS
  • ONLINE EXCLUSIVE PRICE
    214.00 EUR
    202.21 EUR / Pack of 1
    Save 11.79 EUR
    5% Off
Estimated Shipment: 12-03-2027
Wire shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications
Specifications
Product Type Wire Shelf
Material Stainless Steel
Depth (English) 18 in.
Depth (Metric) 457 mm
Dimensions (L x W) 45.7 x 61 cm (18 x 24 in.)
For Use With (Equipment) Metro Super Erecta Shelving System
Includes Plastic sleeves and wedges
Length (English) 24 in.
Length (Metric) 61 cm
Width (English) 18 in.
Width (Metric) 45.7 cm