Learn More
Metro™ Super Erecta™ Swedged Posts
Product Code. 15996691
Change view
Click to view available options
Height (English):
33 in.
54 in.
63 in.
Length (English):
33 in.
54 in.
63 in.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (English) Length (English) Unit Size
15996691 33 in. 33 in. Each
15946711 54 in. 54 in. Each
15976741 63 in. 63 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15996691 Supplier Metro™ Supplier No. 33UPSSW
  • ONLINE EXCLUSIVE PRICE
    59.45 EUR
    51.23 EUR / Pack of 1
    Save 8.22 EUR
    13% Off
Estimated Shipment: 25-03-2027
For cart-washable and autoclave applications.
  • Each Type 304 Stainless Steel post has an aluminum cap swedged into the top
  • For use with standard stem casters
Specifications
Product Type Mobile Shelving Post
For Use With (Application) Autoclavable and Cart Washable Applications
For Use With (Equipment) Metro™ Super Erecta™ stem casters
Height (English) 33 in.
Height (Metric) 83.82 cm
Length (English) 33 in.
Length (Metric) 87.5 cm
Material Stainless Steel