Learn More
Metro™ Super Erecta™ Solid Shelving Accessory, Shelf Divider
Product Code. 15996901
Change view
Click to view available options
Width (English):
14 in.
24 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Unit Size
15996901 24 in. Each
15976901 14 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15996901 Supplier Metro™ Supplier No. DD24FS
  • ONLINE EXCLUSIVE PRICE
    77.20 EUR
    67.18 EUR / Pack of 1
    Save 10.02 EUR
    12% Off
Estimated Shipment: 25-03-2027
Shelf dividers help with organization.
  • Stainless steel
  • 8 in. high
  • Attach with spring clips (included)
Specifications
Product Type Shelf Divider for Wire Shelves
For Use With (Equipment) Metro™ Super Erecta™ shelving
Height (English) 8 in.
Height (Metric) 20.3 cm
Width (English) 24 in.
Width (Metric) 61 cm
Material Stainless Steel