Learn More
Metro™ Starsys™ Cart Accessories
Product Code. 15927251
Change view
Click to view available options
Product Type:
Drawer Pull
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15927251 Drawer Pull Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15927251 Supplier Metro™ Supplier No. SXRDPBK
  • ONLINE EXCLUSIVE PRICE
    9.50 EUR
    5.96 EUR / Pack of 1
    Save 3.54 EUR
    37% Off
Estimated Shipment: 29-01-2027
Parts for use with Starsys carts
  • Black
Specifications
Product Type Drawer Pull
Color Black
For Use With (Equipment) Metro Starsys cart