Learn More
Metro™ Cover Holder for Super Erecta Wire Shelves
Product Code. 15946971
Change view
Click to view available options
Dimensions (L x W x H):
18.25 x 18.5 x 4 in. (46.4 x 47 x 10.16 cm)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
15946971 18.25 x 18.5 x 4 in. (46.4 x 47 x 10.16 cm) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15946971 Supplier Metro™ Supplier No. LARCH
  • ONLINE EXCLUSIVE PRICE
    106.00 EUR
    90.34 EUR / Pack of 1
    Save 15.66 EUR
    14% Off
Estimated Shipment: 02-04-2027
Accessory for Super Erecta Wire Shelf
  • Holds tote box covers
  • Stainless steel wire
  • 18.25L x 18.5W x 4 in. H;
  • 6 lbs.
Specifications
Product Type Cover Holder
Material Stainless Steel
Dimensions (L x W x H) 18.25 x 18.5 x 4 in. (46.4 x 47 x 10.16 cm)
For Use With (Equipment) Metro™ Super Erecta™ shelving system
Height (English) 4 in.
Height (Metric) 10.16 cm
Length (English) 18.25 in.
Length (Metric) 46.4 cm
Weight (English) 6 lb.
Weight (Metric) 2.7 kg
Width (English) 18.5 in.
Width (Metric) 47 cm