missing translation for 'onlineSavingsMsg'
Learn More

Metolazone, MP Biomedicals™

Product Code. 11472972 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472972 100 mg 100mg
11462972 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472972 Supplier MP Biomedicals Supplier No. 0215361780

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diuretic; anti-hypertensive. Moderately soluble in water; soluble in ethanol or DMSO (Store at 2° to 8°C)

Chemical Identifiers

CAS 17560-51-9
Molecular Formula C16H16ClN3O3S
Molecular Weight (g/mol) 365.83
MDL Number MFCD00069304
InChI Key AQCHWTWZEMGIFD-UHFFFAOYNA-N
Synonym metolazone, zaroxolyn, diulo, mykrox, microx, metalozone, metenix, oldren, metolazona, metolazonum
PubChem CID 4170
ChEBI CHEBI:64354
IUPAC Name 7-chloro-2-methyl-3-(2-methylphenyl)-4-oxo-1,2,3,4-tetrahydroquinazoline-6-sulfonamide
SMILES CC1NC2=CC(Cl)=C(C=C2C(=O)N1C1=CC=CC=C1C)S(N)(=O)=O

Specifications

Color White
Quantity 100 mg
Formula Weight 365.83
Physical Form Solid
Chemical Name or Material Metolazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.