Learn More
Metolachlor-ethane sulfonic acid (ESA) sodium 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30317174
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317174 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30317174 Supplier Dr. Ehrenstorfer Supplier No. DREA15171100AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Metolachlor ethane sulfonic acid (ESA) sodium salt
CAS 947601-85-6
Analyte or Component Names Metolachlor ethane sulfonic acid (ESA) sodium salt
Shipping Condition Room Temperature
Molecular Formula C15 H22 N O5 S . Na
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Metolachlor ethane sulfonic acid (ESA) sodium salt
InChI Formula InChI=1S/C15H23NO5S.Na/c1-5-13-8-6-7-11(2)15(13)16(12(3)9-21-4)14(17)10-22(18,19)20;/h6-8,12H,5,9-10H2,1-4H3,(H,18,19,20);/q;+1/p-1
SMILES [Na+].CCc1cccc(C)c1N(C(C)COC)C(=O)CS(=O)(=O)[O-]
IUPAC Name sodium;2-[2-ethyl-N-(1-methoxypropan-2-yl)-6-methylanilino]-2-oxoethanesulfonate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 351.39
Quantity 1 mL
Formula Weight 351.1116 g/mol