missing translation for 'onlineSavingsMsg'
Learn More

Metipranolol hydrochloride, MedChemExpress

Product Code. 16473077
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473077 50 mg 50mg
16467086 100 mg 100mg
16457028 10 mg 10mg
16419577 5 mg 5mg
16413526 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473077 Supplier Medchem Express Supplier No. HY16316
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Metipranolol hydrochloride is a non-selective β adrenergic receptor blocking agent.

Metipranolol hydrochloride is a non-selective β adrenergic receptor blocking agent.

Specifications

Chemical Name or Material Metipranolol hydrochloride
CAS 36592-77-5
Color White
Purity Grade Notes Research
Molecular Formula C17H28ClNO4
Quantity 50 mg
Solubility Information H2O : ≥ 100 mg/mL (289.13 mM) ∣DMSO : 100 mg/mL (289.13 mM; Need ultrasonic)
SMILES CC1=C(OC(C)=O)C(C)=C(C)C(OCC(O)CNC(C)C)=C1.[H]Cl
Molecular Weight (g/mol) 345.86
Formula Weight 345.86
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.