missing translation for 'onlineSavingsMsg'
Learn More

Methylglyoxal, MP Biomedicals™

Product Code. 11494781 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
Unit Size:
100mL
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494781 250 mL 250mL
11474781 25 mL 25mL
11484781 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494781 Supplier MP Biomedicals Supplier No. 0215555883
  • ONLINE EXCLUSIVE PRICE
    307.00 EUR
    291.90 EUR / 250mL
    Save 15.10 EUR
    4% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 78-98-8
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00006960
InChI Key AIJULSRZWUXGPQ-UHFFFAOYSA-N
Synonym methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
PubChem CID 880
ChEBI CHEBI:17158
IUPAC Name 2-oxopropanal
SMILES CC(=O)C=O

Specifications

Color Colorless
Density 1.17g/mL
Quantity 250 mL
Formula Weight 72.06
Physical Form Liquid
Chemical Name or Material Methylglyoxal, Molecular biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.