missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Methylergometrine maleate
GREENER_CHOICE

Product Code. 30001099
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
This item is not returnable. View return policy

Product Code. 30001099

Brand: Tocris Bioscience™ 0549/250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active metabolite of methysergide (Cat. No. 1064)

Active metabolite of methysergide (Cat. No. 1064); uterotonic agent and oxytocic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57432-61-8
Molecular Formula C24H29N3O6

Specifications

Quantity 250 mg
Formula Weight 455.51
Purity >98%
Molecular Weight (g/mol) 455.511
Chemical Name or Material Methylergometrine maleate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.