Learn More
Methylenesuccinic Acid, Sigma-Aldrich™
Product Code. 15879180
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15879180 100 g 100g
15816390 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15879180 Supplier Sigma Aldrich Supplier No. 8048120100
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 97-65-4
Specifications
Melting Point 163°C to 168.4°C
Density 0.949 g/cm3 at 25°C
Molecular Formula HOOCCH2C(=CH2)COOH
MDL Number MFCD00004260
Quantity 100 g
Synonym Methylenesuccinic acid, Itaconic acid, Methylenesuccinic acid
Molecular Weight (g/mol) 130.1
Grade Research