missing translation for 'onlineSavingsMsg'
Learn More

Methylenediurea, TRC

Product Code. 30473608
Change view
Click to view available options
Quantity:
3 mg
6 mg
30 mg
Unit Size:
30mg
3mg
6mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473608 6 mg 6mg
30469291 3 mg 3mg
30464520 30 mg 30mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30473608 Supplier Toronto Research Chemicals Supplier No. TRCM3277536MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13547-17-6
Molecular Formula C3 H8 N4 O2
Molecular Weight (g/mol) 132.12
Synonym Urea, N,N''-methylenebis-, Urea, 1,1'-methylenedi- (6CI,7CI,8CI), N,N''-Methylenebis[urea], 1,1'-Methylenebisurea, 1,1'-Methylenediurea, Diureidomethane, M 2U, Methylenebisurea, Methylenediurea, NSC 36621, NSC 643493
IUPAC Name (carbamoylamino)methylurea
SMILES NC(=O)NCNC(=O)N

Specifications

Quantity 6 mg
InChI Formula InChI=1S/C3H8N4O2/c4-2(8)6-1-7-3(5)9/h1H2,(H3,4,6,8)(H3,5,7,9)
Formula Weight 132.0647
Chemical Name or Material N,N''-Methylenebisurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.