missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue (C.I. 52015), Supelco™

Product Code. 15856130
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15856130 100 g 100g
15846130 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15856130 Supplier Supelco Supplier No. 1592700100
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7220-79-3

Specifications

Melting Point 190°C (dec.) (lit.)
MDL Number MFCD00012111
Synonym Methylene Blue trihydrate, Methylthioninium chloride, 3,7-Bis(dimethylamino)phenothiazinium chloride, Tetramethylthionine chloride, Methylene Blue trihydrate
Quantity 100 g
Molecular Weight (g/mol) 373.9
Grade Research
Chemical Name or Material Methylene blue (C.I. 52015)
compliance-icons
Product Identifier
  • Methylene blue (C.I. 52015) Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Methylthioninium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.