missing translation for 'onlineSavingsMsg'
Learn More

Methyle2-(2,5-dimethyl-1,3-thiazol-4-yl)acetate, 97%, Thermo Scientific™

Product Code. p-2705134
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10753853

Brand: Thermo Scientific Maybridge TL01020DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306937-37-1
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.241
MDL Number MFCD02677686
InChI Key AUEJOXGQRQYLDJ-UHFFFAOYSA-N
Synonym methyl 2-2,5-dimethyl-1,3-thiazol-4-yl acetate, methyl 2-2,5-dimethylthiazol-4-yl acetate, methyl 2,5-dimethyl-1,3-thiazol-4-yl acetate, methyl 2-dimethyl-1,3-thiazol-4-yl acetate, acmc-1agna, 4-thiazoleaceticacid, 2,5-dimethyl-, methyl ester, 2,5-dimethyl-thiazol-4-yl-acetic acid methyl ester, 2-2,5-dimethyl-4-thiazolyl acetic acid methyl ester, methyl 2-2,5-dimethyl-1,3-thiazol-4-yl ethanoate
PubChem CID 2801571
IUPAC Name methyl 2-(2,5-dimethyl-1,3-thiazol-4-yl)acetate
SMILES CC1=C(N=C(S1)C)CC(=O)OC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 185.25
Percent Purity 97%
Chemical Name or Material Methyle2-(2, 5-dimethyl-1, 3-thiazol-4-yl)acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.