accessibility menu, dialog, popup

UserName

CAS RN 306937-37-1

CH3NSH3COOCH3

CAS RN 306937-37-1

IUPAC Name: methyl 2-(2,5-dimethyl-1,3-thiazol-4-yl)acetate
Synonyms: methyl 2-(2,5-dimethyl-1,3-thiazol-4-yl)acetate
Molecular Weight (g/mol): 185.24
Molecular Formula: C8H11NO2S
InChi Key: AUEJOXGQRQYLDJ-UHFFFAOYSA-N
SMILES: COC(=O)CC1=C(C)SC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results