Learn More
Methylbenzethonium Chloride, TRC
Product Code. 17921205
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17921205 50 mg 50mg
17990695 25 mg 25mg
17953785 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17921205 Supplier Toronto Research Chemicals Supplier No. TRCM26504550MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25155-18-4
Molecular Formula C28 H44 N O2 . Cl
Molecular Weight (g/mol) 462.11
Synonym Methylbenzethonium Chloride, Benzenemethanaminium, N,N-dimethyl-N-[2-[2-[methyl-4-(1,1,3,3-tetramethylbutyl)phenoxy]ethoxy]ethyl]-, chloride (1:1), Ammonium, benzyldimethyl[2-[2-[[4-(1,1,3,3-tetramethylbutyl)tolyl]oxy]ethoxy]ethyl]-, chloride (8CI), Benzenemethanaminium, N,N-dimethyl-N-[2-[2-[methyl-4-(1,1,3,3-tetramethylbutyl)phenoxy]ethoxy]ethyl]-, chloride (9CI), Benzyldimethyl[2-[2-[[4-(1,1,3,3-tetramethylbutyl)tolyl]oxy]ethoxy]ethyl]ammonium chloride (7CI), Methylbenzethonium chloride (6CI), 2-(2-(p-(Diisobutyl)cresoxy)ethoxy)ethyldimethylbenzylammonium chloride, Diaparene chloride, Hyamine 10X
IUPAC Name benzyl-dimethyl-[2-[2-[2-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES [Cl-].Cc1cc(ccc1OCCOCC[N+](C)(C)Cc2ccccc2)C(C)(C)CC(C)(C)C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C28H44NO2.ClH/c1-23-20-25(28(5,6)22-27(2,3)4)14-15-26(23)31-19-18-30-17-16-29(7,8)21-24-12-10-9-11-13-24;/h9-15,20H,16-19,21-22H2,1-8H3;1H/q+1;/p-1
Formula Weight 461.3061
Chemical Name or Material Methylbenzethonium Chloride