missing translation for 'onlineSavingsMsg'
Learn More

Methyl Vinyl Ketone, Sigma-Aldrich™

Product Code. 15856400
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15856400 250 mL 250mL
15846400 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15856400 Supplier Sigma-Aldrich Supplier No. 8059010250
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stabilized for synthesis

Methyl vinyl ketone (MVK) is an organic compound that belongs to the class of enones. It is a useful synthetic reagent in organic synthesis for the preparation of various organic compounds. MVK is a 3-oxobutyl synthon, which undergoes a 1,4 conjugate addition (Michael addition) reaction. MVK is susceptible to anionic polymerization, therefore it is typically stored with hydroquinone, which inhibits polymerization.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-94-4

Specifications

Density 0.85 g/cm3 at 20°C
Boiling Point 80°C to 81°C/1013 hPa
MDL Number MFCD00008777
Quantity 250 mL
Synonym Methyl vinyl ketone, 1-Buten-3-one, MVK, Vinyl methyl ketone, Methyl vinyl ketone
Molecular Weight (g/mol) 70.09
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.