missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-(+)-3-hydroxy-2-methylpropionate, 98%

Product Code. p-7059373
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420830 2 g 2g
11430830 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420830 Supplier Thermo Scientific Alfa Aesar Supplier No. 044483.04
Add to basket
This item is not returnable. View return policy

CAS: 80657-57-4 | C5H10O3 | 118.13 g/mol

Methyl (S)-(+)-3-hydroxy-2-methylpropionate is used as a starting material for the synthesis of dictyostatin, discodermolide and spongidepsin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80657-57-4
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00064520
InChI Key ATCCIZURPPEVIZ-UHFFFAOYNA-N
Synonym s-methyl 3-hydroxy-2-methylpropanoate, methyl s-+-3-hydroxy-2-methylpropionate, s-+-3-hydroxy-2-methylpropionic acid methyl ester, methyl 2s-3-hydroxy-2-methylpropanoate, 2-methyl-3-hydroxypropanoic acid, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2s, s-+-methyl 3-hydroxy-2-methylpropionate, s-3-hydroxy-2-methylpropionic acid methyl ester, +-methyl l-beta-hydroxyisobutyrate, methyl 3-hydroxy-2-methylpropanoate #
PubChem CID 10192563
IUPAC Name methyl (2S)-3-hydroxy-2-methylpropanoate
SMILES COC(=O)C(C)CO

Specifications

Color Colorless
Density 1.07 g/mL
Boiling Point 74°C (10 mmHg)
Flash Point 81°C (178°F)
Refractive Index 1.425
Quantity 2 g
Beilstein 1720830
Solubility Information Insoluble in water. Soluble in methanol.
Optical Rotation +27° (c=4 in methanol)
Formula Weight 118.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl (S)- (+)-3-hydroxy-2-methylpropionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.