missing translation for 'onlineSavingsMsg'
Learn More

Methyl Pyrophosphate Sodium Salt (~90%), TRC

Product Code. 30481087
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481087 10 mg 10mg
30479870 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481087 Supplier Toronto Research Chemicals Supplier No. TRCM22772510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Pyrophosphate Sodium Salt
Molecular Formula CH6O7P2 . x(Na)
Quantity 10 mg
Synonym Monomethyl Ester Diphosphoric Acid, Sodium Salt
InChI Formula InChI=1S/CH6O7P2.3Na/c1-7-10(5,6)8-9(2,3)4;;;/h1H3,(H,5,6)(H2,2,3,4);;;/q;3*+1/p-3
SMILES O=P([O-])(OP([O-])([O-])=O)OC.[Na+].[Na+].[Na+]
IUPAC Name sodium methyl diphosphate
Molecular Weight (g/mol) 257.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.