missing translation for 'onlineSavingsMsg'
Learn More

Methyl Propiolate, MP Biomedicals

Product Code. 11432586 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432586 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11432586 Supplier MP Biomedicals Supplier No. 0521548210

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 922-67-8
Molecular Formula C4H4O2
Quantity 10 g
Synonym methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester
InChI Key IMAKHNTVDGLIRY-UHFFFAOYSA-N
SMILES COC(=O)C#C
IUPAC Name methyl prop-2-ynoate
Molecular Weight (g/mol) 84.074
PubChem CID 13536
Formula Weight 84.07
Chemical Name or Material Methyl Propiolate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.