Learn More
Methyl Levulinate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Levulinate |
| CAS | 624-45-3 |
| Molecular Formula | C6H10O3 |
| Quantity | 100 g |
| Synonym | 4-Oxopentanoic Acid Methyl Ester, 4-Oxovaleric Acid Methyl Ester, Methyl 4-Oxopentanoate, Methyl 4-Oxovalerate, Methyl Levulate, Methyl beta-Acetylpropionate, NSC 24874 |
| InChI Formula | InChI=1S/C6H10O3/c1-5(7)3-4-6(8)9-2/h3-4H2,1-2H3 |
| SMILES | COC(=O)CCC(=O)C |
| IUPAC Name | methyl 4-oxopentanoate |
| Molecular Weight (g/mol) | 130.14 |
| Formula Weight | 130.063 |